O-Benzyl-N-(tert-butoxycarbonyl)-D-serine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B1863-1G
B1863-5G
TCB1863-1GEA
109.02
CAD
TCB1863-1G
TCB1863-005G
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine ≥98.0% (by HPLC, titration analysis)
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine
CAS Number: 47173-80-8
MDL Number: MFCD00038248
Molecular Formula: C15H21NO5
Molecular Weight: 295.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 60
Specific rotation [a]20/D: -20 deg (C=2, EtOH(80vol%))
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-O-benzyl-D-serine, Boc-D-Ser(Bzl)-OH
Formula:
C₁₅H₂₁NO₅ Melting Pt: 60 °C |
MDL Number:
MFCD00038248 CAS Number: 47173-80-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...