Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine ≥98.0%

Supplier: TCI America

Synonyms: Nε-Boc-Nɑ-Cbz-L-lysine, Z-Lys(Boc)-OH

B1862-25G B1862-5G
TCB1862-25GEA 718.9 CAD
TCB1862-25G TCB1862-005G
Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine ≥98.0%
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-carbobenzoxy-L-lysine

CAS Number: 2389-60-8
MDL Number: MFCD00062271
Molecular Formula: C19H28N2O6
Molecular Weight: 380.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 62
Specific rotation [a]20/D: -6 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nepsilon-Boc-Nalpha-Cbz-L-lysine, Z-Lys(Boc)-OH

Formula: C₁₉H₂₈N₂O₆
MW: 380.44 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00062271
CAS Number: 2389-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6599874e17a8d1edb2d02e0aea847dced7b0fa21&vid=6599874e17a8d1edb2d02e0aea847dced7b0fa21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR