4-Bromobenzylamine hydrochloride ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B1841-25G B1841-5G
TCB1841-25GEA 748.92 CAD
TCB1841-25G TCB1841-005G
4-Bromobenzylamine hydrochloride ≥97.0% (by total nitrogen and titration analysis)
4-Bromobenzylamine hydrochloride

CAS Number: 26177-44-6
MDL Number: MFCD00012860
Molecular Formula: C7H8BrN
Molecular Weight: 222.51
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal

Formula: C₇H₉BrClN
MDL Number: MFCD00012860
CAS Number: 26177-44-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bed471ec71f39255111820d441fbab562f7c873&vid=3bed471ec71f39255111820d441fbab562f7c873&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR