2-Bromobenzylamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1836-25G
B1836-5G
TCB1836-025GEA
358.66
CAD
TCB1836-025G
TCB1836-005G
2-Bromobenzylamine hydrochloride ≥98.0% (by titrimetric analysis)
2-Bromobenzylamine hydrochloride
CAS Number: 5465-63-4
MDL Number: MFCD00012853
Molecular Formula: C7H8BrN
Molecular Weight: 222.51
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₇H₉BrClN MW: 222.52 g/mol |
MDL Number:
MFCD00012853 CAS Number: 5465-63-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...