2,2-Bis(4-glycidyloxyphenyl)propane ≥85.0%

Supplier: TCI America

Synonyms: Bisphenol A Diglycidyl Ether, 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, DER 332, BADGE, D.E.R. 332 epoxy resin

B1796-25G B1796-500G
TCB1796-025GEA 45 CAD
TCB1796-025G TCB1796-500G
2,2-Bis(4-glycidyloxyphenyl)propane ≥85.0%
2,2-Bis(4-glycidyloxyphenyl)propane

CAS Number: 1675-54-3
MDL Number: MFCD00080480
Molecular Formula: C21H24O4
Molecular Weight: 340.42
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 150
Specific Gravity (20/20): 1.17


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, Bisphenol A Diglycidyl Ether

Formula: C₂₁H₂₄O₄
MW: 340.42 g/mol
Boiling Pt: 210 °C
Density: 1.17 (20 °C)
Flash Pt: 150 °C
MDL Number: MFCD00080480
CAS Number: 1675-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e49bb67d930d05c62c5a8e61a85da2c687f9dd32&vid=e49bb67d930d05c62c5a8e61a85da2c687f9dd32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR