Diethyl (2-bromoethyl)phosphonate ≥95.0%
Supplier: TCI America
B1781-25G
B1781-5G
TCB1781-025GEA
456.62
CAD
TCB1781-025G
TCB1781-5G
Diethyl (2-bromoethyl)phosphonate ≥95.0%
Diethyl (2-bromoethyl)phosphonate
CAS Number: 5324-30-1
MDL Number: MFCD00000239
Molecular Formula: C6H14BrO3P
Molecular Weight: 245.05
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 110
Specific Gravity (20/20): 1.38
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 2-Bromoethylphosphonic Acid Diethyl Ester
Formula:
C₆H₁₄BrO₃P MW: 245.06 g/mol Boiling Pt: 115 °C Density: 1.38 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00000239 CAS Number: 5324-30-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...