(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥98.0%
Supplier: TCI America
Synonyms:
PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
B1774-5G
B1774-25G
TCB1774-005GEA
130.37
CAD
TCB1774-005G
TCB1774-25G
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥98.0%
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
[Coupling Reagent for Peptide]
CAS Number: 128625-52-5
MDL Number: MFCD00077411
Molecular Formula: C18H28F6N6OP2
Molecular Weight: 520.40
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₁₈H₂₈F₆N₆OP₂ MW: 520.4 g/mol |
MDL Number:
MFCD00077411 CAS Number: 128625-52-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...