1H-Benzimidazole-5-carboxylic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid
B1763-25G
B1763-5G
TCB1763-025GEA
436.08
CAD
TCB1763-025G
TCB1763-005G
1H-Benzimidazole-5-carboxylic acid ≥97.0% (by titrimetric analysis)
1H-Benzimidazole-5-carboxylic acid
CAS Number: 15788-16-6
MDL Number: MFCD00011555
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Other Product Names / Synonyms: 5-Carboxybenzimidazole
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol |
MDL Number:
MFCD00011555 CAS Number: 15788-16-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...