(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine ≥95.0%

Supplier: TCI America

Synonyms: (S)-(-)-3-Boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, (S)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde

B1759-1G B1759-5G
TCB1759-001GEA 241.27 CAD
TCB1759-001G TCB1759-005G
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine ≥95.0%
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine

CAS Number: 102308-32-7
MDL Number: MFCD00209557
Molecular Formula: C11H19NO4
Molecular Weight: 229.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 107
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: -91.5 deg (C=1, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-3-Boc-4-formyl-2,2-dimethyl-1,3-oxazolidine

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Boiling Pt: 97 °C
Density: 1.06 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00209557
CAS Number: 102308-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=751638f69a32cdbfc9e3374bace8f238284542cf&vid=751638f69a32cdbfc9e3374bace8f238284542cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR