N,N'-Bis(2-hydroxyethyl)oxamide ≥98.0%

Supplier: TCI America

Synonyms: N,N'-Oxalylbis(ethanolamine), N1,N2-Bis(2-hydroxyethyl)oxalamide

B1718-5G B1718-25G
TCB1718-5GEA 58.46 CAD
TCB1718-5G TCB1718-025G
N,N'-Bis(2-hydroxyethyl)oxamide ≥98.0%
N,N'-Bis(2-hydroxyethyl)oxamide

CAS Number: 1871-89-2
MDL Number: MFCD00020559
Molecular Formula: C6H12N2O4
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: N,N'-Oxalylbis(ethanolamine)

Formula: C₆H₁₂N₂O₄
Melting Pt: 169 °C
MDL Number: MFCD00020559
CAS Number: 1871-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1d1c08d1db2ece00073a842cde9f2a872fa4b39&vid=c1d1c08d1db2ece00073a842cde9f2a872fa4b39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR