Sodium-3-bromopropanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromopropanesulfonic acid Sodium Salt, 3-Bromopropanesulphonic acid Sodium Salt

B1707-25G B1707-100G
TCB1707-025GEA 199.08 CAD
TCB1707-025G TCB1707-100G
Sodium-3-bromopropanesulfonate ≥98.0%
Sodium-3-bromopropanesulfonate

CAS Number: 55788-44-8
MDL Number: MFCD00024999
Molecular Formula: C3H7BrO3S
Molecular Weight: 225.03
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3-Bromopropanesulfonic Acid Sodium Salt

Formula: C₃H₇BrO₃S
MW: 225.03 g/mol
MDL Number: MFCD00024999
CAS Number: 55788-44-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=077fd9287654cd551c754d85bc5d4b014c14bac0&vid=077fd9287654cd551c754d85bc5d4b014c14bac0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR