N-tert-Butyl-ɑ-phenylnitrone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B1701-5G B1701-1G
TCB1701-005GEA 346.02 CAD
TCB1701-005G TCB1701-001G
N-tert-Butyl-ɑ-phenylnitrone ≥98.0% (by HPLC, titration analysis)
N-tert-Butyl-alpha-phenylnitrone

CAS Number: 3376-24-7
MDL Number: MFCD00008799
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 74


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: N-Benzylidene-tert-butylamine N-Oxide, PBN

Formula: C₁₁H₁₅NO
Melting Pt: 74 °C
MDL Number: MFCD00008799
CAS Number: 3376-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=237f7fcd897c18e8aba0ef14b092ed0d27ef2c61&vid=237f7fcd897c18e8aba0ef14b092ed0d27ef2c61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR