Bis(8-quinolinolato)zinc(II) hydrate ≥93.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Zinc 8-hydroxyquinolinate, Bis(8-hydroxyquinolinato)zinc(II), Zinc(II) Bis(8-hydroxyquinolinate)

B1678-25G B1678-5G
TCB1678-025GEA 357.08 CAD
TCB1678-025G TCB1678-5G
Bis(8-quinolinolato)zinc(II) hydrate ≥93.0% (by titrimetric analysis)
Bis(8-quinolinolato)zinc(II) hydrate

CAS Number: 13978-85-3
MDL Number: MFCD00065291
Molecular Formula: C18H12N2O2Zn
Molecular Weight: 353.69
Purity/Analysis Method: >93.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: Bis(8-hydroxyquinolinato)zinc(II), Zinc(II) Bis(8-hydroxyquinolinate)

Formula: C₁₈H₁₂N₂O₂Zn
MW: 353.69 g/mol
MDL Number: MFCD00065291
CAS Number: 13978-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2afa6ae267163eee77338e2824e31fb71687b63&vid=f2afa6ae267163eee77338e2824e31fb71687b63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR