Nε-(tert-butoxycarbonyl)-L-lysine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Nε-Boc-L-lysine, H-Lys(Boc)-OH
B1670-5G
B1670-1G
TCB1670-005GEA
323.16
CAD
TCB1670-005G
TCB1670-001G
Nε-(tert-butoxycarbonyl)-L-lysine ≥97.0% (by titrimetric analysis)
Nepsilon-(tert-butoxycarbonyl)-L-lysine
CAS Number: 2418-95-3
MDL Number: MFCD00037221
Molecular Formula: C11H22N2O4
Molecular Weight: 246.31
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: 18 deg (C=1, AcOH)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: Nepsilon-Boc-L-lysine, H-Lys(Boc)-OH
Formula:
C₁₁H₂₂N₂O₄ MW: 246.31 g/mol |
MDL Number:
MFCD00037221 CAS Number: 2418-95-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...