HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)

B1657-5G B1657-25G B1657-100G
TCB1657-005GEA 91.97 CAD
TCB1657-005G TCB1657-025G TCB1657-100G
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC)
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

[Coupling Reagent for Peptide]
CAS Number: 94790-37-1
MDL Number: MFCD00075445
Molecular Formula: C11H16F6N5OP
Molecular Weight: 379.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: HBTU, N,N,N',N'-Tetramethyl-O-(benzotriazol-1-yl)uronium Hexafluorophosphate

Formula: C₁₁H₁₆F₆N₅OP
MW: 379.2 g/mol
MDL Number: MFCD00075445
CAS Number: 94790-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11e55ef0528235d184569fd473a835967a9b5003&vid=e0bb7900d5d37f76bb9927b2334502591fc93414&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR