2-(Benzyloxy)benzaldehyde ≥98.0%
Supplier: TCI America
B1653-5G
B1653-25G
TCB1653-5GEA
41.08
CAD
TCB1653-5G
TCB1653-25G
2-(Benzyloxy)benzaldehyde ≥98.0%
2-(Benzyloxy)benzaldehyde
CAS Number: 5896-17-3
MDL Number: MFCD00016583
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Specific Gravity (20/20): 1.15
Formula:
C₁₄H₁₂O₂ Boiling Pt: 208 °C Density: 1.15 (20 °C) |
MDL Number:
MFCD00016583 CAS Number: 5896-17-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...