(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98.0% (by HPLC)

Supplier: TCI America

B1651-5G B1651-25G B1651-100G
TCB1651-005GEA 94.96 CAD
TCB1651-005G TCB1651-25G TCB1651-100G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98.0% (by HPLC)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)

CAS Number: 56602-33-6
MDL Number: MFCD00011948
Molecular Formula: C12H22F6N6OP2
Molecular Weight: 442.29
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Lambda max.: 255 nm (CH3CN)


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: BOP, Castro's Reagent

Formula: C₁₂H₂₂F₆N₆OP₂
MDL Number: MFCD00011948
CAS Number: 56602-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c458561009df8acf15278cc27230972959580a09&vid=735da205a41943c26d2fcb68e7994d57c934e9be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR