(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid ≥98.0%
Supplier: TCI America
Synonyms:
Boc-Gln-OH, N(α)-Boc-L-glutamine, Nalpha-Boc-L-glutamine
B1649-10G
B1649-25G
TCB1649-010GEA
107.44
CAD
TCB1649-010G
TCB1649-25G
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid ≥98.0%
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid
CAS Number: 13726-85-7
MDL Number: MFCD00065571
Molecular Formula: C10H18N2O5
Molecular Weight: 246.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -4 deg (C=2, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-L-glutamine, Boc-Gln-OH
Formula:
C10H18N2O5 MW: 246.3 g/mol |
MDL Number:
MFCD00065571 CAS Number: 13726-85-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...