N-Boc-L-serine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
B1637-5G
B1637-25G
TCB1637-005GEA
42.66
CAD
TCB1637-005G
TCB1637-025G
N-Boc-L-serine ≥97.0% (by titrimetric analysis)
N-Boc-L-serine
CAS Number: 3262-72-4
MDL Number: MFCD00037243
Molecular Formula: C8H15NO5
Molecular Weight: 205.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: -4 deg (C=1, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-serine, Boc-Ser-OH
Formula:
C₈H₁₅NO₅ |
MDL Number:
MFCD00037243 CAS Number: 3262-72-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...