N(α)-Boc-L-histidine ≥98.0%

Supplier: TCI America

Synonyms: Boc-His-OH

B1634-5G B1634-25G
TCB1634-005GEA 142.2 CAD
TCB1634-005G TCB1634-025G
N(α)-Boc-L-histidine ≥98.0%
N(α)-Boc-L-histidine

CAS Number: 17791-52-5
MDL Number: MFCD00065576
Molecular Formula: C11H17N3O4
Molecular Weight: 255.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 26 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-L-histidine, Boc-His-OH

Formula: C₁₁H₁₇N₃O₄
MDL Number: MFCD00065576
CAS Number: 17791-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1f11c297b8fd2557b21ac315d673ffd9db9530a&vid=e1f11c297b8fd2557b21ac315d673ffd9db9530a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR