Benzyl-2-naphthyl ether ≥98.0%

Supplier: TCI America

Synonyms: Benzyl 2-Naphthyl Ether, 2-Benzyloxynaphthalene

B1619-500G B1619-25G
TCB1619-500GEA 135.56 CAD
TCB1619-500G TCB1619-025G
Benzyl-2-naphthyl ether ≥98.0%
Benzyl-2-naphthyl ether

CAS Number: 613-62-7
MDL Number: MFCD00089092
Molecular Formula: C17H14O
Molecular Weight: 234.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 2-Benzyloxynaphthalene

Formula: C₁₇H₁₄O
MW: 234.30 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00089092
CAS Number: 613-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=671b766536983f819a49da86079ab6b6bb7d5582&vid=671b766536983f819a49da86079ab6b6bb7d5582&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR