1-Bromo-2-(bromomethyl)naphthalene ≥97.0%
Supplier: TCI America
B1607-1G
B1607-5G
B1607-25G
TCB1607-001GEA
86.9
CAD
TCB1607-001G
TCB1607-005G
TCB1607-25G
1-Bromo-2-(bromomethyl)naphthalene ≥97.0%
1-Bromo-2-(bromomethyl)naphthalene
CAS Number: 37763-43-2
MDL Number: MFCD00046369
Molecular Formula: C11H8Br2
Molecular Weight: 299.99
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 108
Other Product Names / Synonyms: 2-(Bromomethyl)-1-bromonaphthalene
Formula:
C₁₁H₈Br₂ Melting Pt: 108 °C |
MDL Number:
MFCD00046369 CAS Number: 37763-43-2 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...