4-Benzylamino-1-butanol ≥96.0%

Supplier: TCI America

Synonyms: N-(4-Hydroxybutyl)benzylamine

B1599-5G B1599-25G
TCB1599-5GEA 67.78 CAD
TCB1599-5G TCB1599-025G
4-Benzylamino-1-butanol ≥96.0%
4-Benzylamino-1-butanol

CAS Number: 59578-63-1
MDL Number: MFCD00191609
Molecular Formula: C11H17NO
Molecular Weight: 179.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: N-(4-Hydroxybutyl)benzylamine

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
Boiling Pt: 140 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00191609
CAS Number: 59578-63-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc816555d7b90b5e184176fcc5baeeb611411357&vid=dc816555d7b90b5e184176fcc5baeeb611411357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR