(S)-(-)-N-Benzyl-α-methylbenzylamine ≥98.0%

Supplier: TCI America

Synonyms: S-(-)-N-Benzyl-1-phenylethylamine, (S)-(-)-N-Benzyl-1-phenylethylamine

B1529-25ML B1529-5ML
TCB1529-25MLEA 172.22 CAD
TCB1529-25ML TCB1529-005ML
(S)-(-)-N-Benzyl-α-methylbenzylamine ≥98.0%
(S)-(-)-N-Benzyl-α-methylbenzylamine

CAS Number: 17480-69-2
MDL Number: MFCD00066325
Molecular Formula: C15H17N
Molecular Weight: 211.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.01
Specific rotation [a]20/D: -39 deg (neat)


Other Product Names / Synonyms: (S)-(-)-N-Benzyl-alpha-methylbenzylamine

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Density: 1.01 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00066325
CAS Number: 17480-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=642a4882ff56340a6ff71f18474493ed3a6d0a73&vid=642a4882ff56340a6ff71f18474493ed3a6d0a73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR