(R)-(+)-N-Benzyl-α-methylbenzylamine ≥98.0%

Supplier: TCI America

Synonyms: (+)-Benzylphenethylamine, (R)-N-Benzyl-1-phenylethanamine, (R)-(+)-N-Benzyl-1-phenylethylamine

B1528-25ML B1528-5ML
TCB1528-25MLEA 155.31 CAD
TCB1528-25ML TCB1528-005ML
(R)-(+)-N-Benzyl-α-methylbenzylamine ≥98.0%
(R)-(+)-N-Benzyl-α-methylbenzylamine

CAS Number: 38235-77-7
MDL Number: MFCD00015010
Molecular Formula: C15H17N
Molecular Weight: 211.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 158
Specific Gravity (20/20): 1.01
Specific rotation [a]20/D: 39 deg (neat)


Other Product Names / Synonyms: (R)-(+)-N-Benzyl-alpha-methylbenzylamine

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Density: 1.01 (20 °C)
Flash Pt: 158 °C
MDL Number: MFCD00015010
CAS Number: 38235-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=851fe6bc5fed0251e98423795d7b5a6b857cfcac&vid=851fe6bc5fed0251e98423795d7b5a6b857cfcac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR