1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98.0%

Supplier: TCI America

Synonyms: 2,2'-(Benzene-1,3-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

B1403-25G B1403-5G
TCB1403-025GEA 469.26 CAD
TCB1403-025G TCB1403-005G
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98.0%
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene

CAS Number: 802-93-7
MDL Number: MFCD00042090
Molecular Formula: C12H6F12O2
Molecular Weight: 410.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 97
Specific Gravity (20/20): 1.66


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Bis(alpha-hydroxyhexafluoroisopropyl)benzene, alpha,alpha,alpha',alpha'-Tetrakis(trifluoromethyl)-1,3-benzenedimethanol

Formula: C₁₂H₆F₁₂O₂
MW: 410.16
Boiling Pt: 100 °C
Density: 1.66 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00042090
CAS Number: 802-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=139770db9889cdd61b1cee72f303cfd73da2e641&vid=139770db9889cdd61b1cee72f303cfd73da2e641&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR