2,2-Bis(4-carboxyphenyl)hexafluoropropane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1401-5G B1401-25G
TCB1401-005GEA 110.6 CAD
TCB1401-005G TCB1401-025G
2,2-Bis(4-carboxyphenyl)hexafluoropropane ≥98.0% (by GC, titration analysis)
2,2-Bis(4-carboxyphenyl)hexafluoropropane

CAS Number: 1171-47-7
MDL Number: MFCD00040931
Molecular Formula: C17H10F6O4
Molecular Weight: 392.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 275


Product Categories: Materials Chemistry, Monomers, Bisphenol AF type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-(Hexafluoroisopropylidene)bis(benzoic Acid)

Formula: C₁₇H₁₀F₆O₄
Melting Pt: 275 °C
MDL Number: MFCD00040931
CAS Number: 1171-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46ea288e039d5ef11e274da061ee25769eb77d9&vid=f46ea288e039d5ef11e274da061ee25769eb77d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR