Bis(dibenzylideneacetone)palladium
Supplier: TCI America
B1374-1G
B1374-5G
TCB1374-001GEA
167.01
CAD
TCB1374-001G
TCB1374-005G
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium
CAS Number: 32005-36-0
MDL Number: MFCD00051942
Molecular Formula: C34H28O2Pd
Molecular Weight: 575.02
Form: Crystal
Color: Deep Yellow Red
Melting point (°C): 150
Other Product Names / Synonyms: Palladium(0) Bis(dibenzylideneacetone)
Formula:
C34H28O2Pd Melting Pt: 150 °C |
MDL Number:
MFCD00051942 CAS Number: 32005-36-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...