Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1367-5G B1367-25G
TCB1367-005GEA 137.46 CAD
TCB1367-005G TCB1367-25G
Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥98.0% (by titrimetric analysis)
Bis[2-carboxyethylgermanium(IV)] sesquioxide

CAS Number: 12758-40-6
MDL Number: MFCD00049319
Molecular Formula: C6H10Ge2O7
Molecular Weight: 339.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 320

Formula: C₆H₁₀Ge₂O₇
Melting Pt: 320 °C
MDL Number: MFCD00049319
CAS Number: 12758-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa6441bf0658749e4054014bf4d9c0712d508e8c&vid=fa6441bf0658749e4054014bf4d9c0712d508e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR