N-Boc-L-valine ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Boc-Val-OH, BOC-L-Valine
B1333-5G
B1333-25G
TCB1333-005GEA
28.44
CAD
TCB1333-005G
TCB1333-025G
N-Boc-L-valine ≥99.0% (by titrimetric analysis)
N-Boc-L-valine
CAS Number: 13734-41-3
MDL Number: MFCD00065605
Molecular Formula: C10H19NO4
Molecular Weight: 217.27
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 78
Specific rotation [a]20/D: -6.5 deg (C=1, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-valine, Boc-Val-OH
Formula:
C₁₀H₁₉NO₄ MW: 217.3 g/mol Melting Pt: 78 °C |
MDL Number:
MFCD00065605 CAS Number: 13734-41-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...