4-Bromo-2,2-diphenylbutanenitrile ≥95.0%

Supplier: TCI America

B1331-25G B1331-250G
TCB1331-025GEA 44.24 CAD
TCB1331-025G TCB1331-250G
4-Bromo-2,2-diphenylbutanenitrile ≥95.0%
4-Bromo-2,2-diphenylbutanenitrile

CAS Number: 39186-58-8
MDL Number: MFCD00001845
Molecular Formula: C16H14BrN
Molecular Weight: 300.20
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 70

Formula: C₁₆H₁₄BrN
Melting Pt: 70 °C
MDL Number: MFCD00001845
CAS Number: 39186-58-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b227c9ab929c377fe6837169657805037260fdc&vid=3b227c9ab929c377fe6837169657805037260fdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR