2-Bromoterephthalic acid ≥98.0%
Supplier: TCI America
B1321-5G
B1321-25G
TCB1321-5GEA
280.92
CAD
TCB1321-5G
TCB1321-25G
2-Bromoterephthalic acid ≥98.0%
2-Bromoterephthalic acid
Formula:
C₈H₅BrO₄ |
MDL Number:
MFCD00002403 CAS Number: 586-35-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...