Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1317-25G
B1317-500G
TCB1317-025GEA
71.1
CAD
TCB1317-025G
TCB1317-500G
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
CAS Number: 1719-83-1
MDL Number: MFCD00082228
Molecular Formula: C12H8O6
Molecular Weight: 248.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₁₂H₈O₆ MW: 248.19 g/mol |
MDL Number:
MFCD00082228 CAS Number: 1719-83-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...