Benzofuroxan ≥98.0%

Supplier: TCI America

B1316-25G B1316-500G
TCB1316-025GEA 82.16 CAD
TCB1316-025G TCB1316-500G
Benzofuroxan ≥98.0%
Benzofuroxan

CAS Number: 480-96-6
MDL Number: MFCD00005807
Molecular Formula: C6H4N2O2
Molecular Weight: 136.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69


Other Product Names / Synonyms: Benzofurazan 1-Oxide, 2,1,3-Benzoxadiazole 1-Oxide

Formula: C₆H₄N₂O₂
Melting Pt: 69 °C
MDL Number: MFCD00005807
CAS Number: 480-96-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78dd75d645c1927479e8dc93ea5a5571cc4e6d3b&vid=78dd75d645c1927479e8dc93ea5a5571cc4e6d3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR