Dimethyl biphenyl-4,4'-dicarboxylate ≥98.0%
Supplier: TCI America
B1309-25G
B1309-5G
TCB1309-025GEA
172.22
CAD
TCB1309-025G
TCB1309-5G
Dimethyl biphenyl-4,4'-dicarboxylate ≥98.0%
Dimethyl biphenyl-4,4'-dicarboxylate
CAS Number: 792-74-5
MDL Number: MFCD00017201
Molecular Formula: C16H14O4
Molecular Weight: 270.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 217
Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Bibenzoic Acid Dimethyl Ester, 4,4'-Biphenyldicarboxylic Acid Dimethyl Ester
Formula:
C₁₆H₁₄O₄ Melting Pt: 217 °C |
MDL Number:
MFCD00017201 CAS Number: 792-74-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...