4,4'-Isopropylidenediphenyl diacetate ≥98.0%
Supplier: TCI America
Synonyms:
Bisphenol A diacetate, 2,2-Bis(4-acetoxyphenyl)propane, 4,4'-Isopropylidenediphenol diacetate
CAS Number: 10192-62-8
MDL Number: MFCD00026194
Molecular Formula: C19H20O4
Molecular Weight: 312.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 92
Product Categories: Materials Chemistry, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bisphenol A Diacetate, 4,4'-Isopropylidenediphenol Diacetate
Formula:
C₁₉H₂₀O₄ Boiling Pt: 215 °C Melting Pt: 92 °C |
MDL Number:
MFCD00026194 CAS Number: 10192-62-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...