5-Bromo-1,3-benzodioxole ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

B1230-25G B1230-5G
TCB1230-025GEA 147.1 CAD
TCB1230-025G TCB1230-5G
5-Bromo-1,3-benzodioxole ≥98.0%
5-Bromo-1,3-benzodioxole

CAS Number: 2635-13-4
MDL Number: MFCD00005821
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 110
Specific Gravity (20/20): 1.68


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 5-Bromo-1,3-benzodioxole, 3,4-Methylenedioxybromobenzene

Formula: C₇H₅BrO₂
Boiling Pt: 228 °C
Density: 1.68 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005821
CAS Number: 2635-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=739b27da5fead3813e02db8c0163a58ff9f440f3&vid=739b27da5fead3813e02db8c0163a58ff9f440f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR