Boc-L-leucine monohydrate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
B1187-5G
B1187-25G
TCB1187-005GEA
44.24
CAD
TCB1187-005G
TCB1187-025G
Boc-L-leucine monohydrate ≥99.0% (by titrimetric analysis)
Boc-L-leucine monohydrate
CAS Number: 13139-15-6
MDL Number: MFCD00066067
Molecular Formula: C11H21NO4
Molecular Weight: 231.29
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-leucine, Boc-Leu-OH
Formula:
C₁₁H₂₁NO₄·H₂O Melting Pt: 87 °C |
MDL Number:
MFCD00065582 CAS Number: 200936-87-4 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity(Neutralization titration) | min. 99.0%(calcd.on anh.substance) |
Melting point | 85.0 to 88.0 deg-C |
Specific rotation [a]20/D | -24.0 to -26.0 deg(C=2, AcOH) |
Solubility in Acetic acid | almost transparency |
Water | 6.0 to 8.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...