Boc-L-isoleucine ≥99.0%

Supplier: TCI America

Synonyms: N-Boc-L-isoleucine, Boc-Ile-OH

B1186-5G B1186-25G
TCB1186-005GEA 53.53 CAD
TCB1186-005G TCB1186-025G
Boc-L-isoleucine ≥99.0%
Boc-L-isoleucine

CAS Number: 13139-16-7
MDL Number: MFCD00038324
Molecular Formula: C11H21NO4
Molecular Weight: 231.29
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 60
Specific rotation [a]20/D: 2.7 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-isoleucine, Boc-Ile-OH

Formula: C₁₁H₂₁NO₄
MW: 231.06 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00038324
CAS Number: 13139-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c233f9266121d9fee21d39791a01202e46fe5bc&vid=8c233f9266121d9fee21d39791a01202e46fe5bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR