N-Boc-glycine (boc-gly-OH) ≥98.0%

Supplier: TCI America

Synonyms: Boc-Gly-OH

B1185-5G B1185-25G
TCB1185-005GEA 38.58 CAD
TCB1185-005G TCB1185-025G
N-Boc-glycine (boc-gly-OH) ≥98.0%
N-Boc-glycine (Boc-Gly-OH)

CAS Number: 4530-20-5
MDL Number: MFCD00002690
Molecular Formula: C7H13NO4
Molecular Weight: 175.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 89


Product Categories: Bioscience, Amino Acids (N-Protected), Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: N-Boc-glycine, Boc-Gly-OH

Formula: C₇H₁₃NO₄
Melting Pt: 89 °C
MDL Number: MFCD00002690
CAS Number: 4530-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bd5ff254d24a7aed78b531a1512e186ea8eb24f&vid=6bd5ff254d24a7aed78b531a1512e186ea8eb24f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR