N-Boc-L-alanine 98,0

Supplier: TCI America

Synonyms: N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH

B1184-5G B1184-25G
TCB1184-005GEA 47.4 CAD
TCB1184-005G TCB1184-025G
N-Boc-L-alanine 98,0
N-Boc-L-alanine

CAS Number: 15761-38-3
MDL Number: MFCD00037225
Molecular Formula: C8H15NO4
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: -25.5 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-alanine, Boc-Ala-OH

Formula: C8H15NO4
MW: 189.21 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00037225
CAS Number: 15761-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa02cc4ace601cf542f02b8a9356d5e16b83b8c9&vid=fa02cc4ace601cf542f02b8a9356d5e16b83b8c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR