3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0%
Supplier: TCI America
Synonyms:
2,2'-(2,4,8,10-Tetraoxaspiro[5.5]undecane-3,9-diyl)bis(2-methylpropan-1-ol), Bis(hydroxypivalaldehyde) Pentaerythritolacetal Cyclic Acetal
CAS Number: 1455-42-1
MDL Number: MFCD00059794
Molecular Formula: C15H28O6
Molecular Weight: 304.38
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 196
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bis(hydroxypivalaldehyde) Pentaerythritolacetal Cyclic Acetal
Formula:
C₁₅H₂₈O₆ MW: 304.38 g/mol Melting Pt: 196 °C |
MDL Number:
MFCD00059794 CAS Number: 1455-42-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...