(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
CAS Number: 35193-64-7
MDL Number: MFCD00010045
Molecular Formula: C20H13O4P
Molecular Weight: 348.29
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: 595 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(+)-2,2'-Dihydroxy-1,1'-binaphthyl Hydrogen Phosphate
Formula:
C₂₀H₁₃O₄P MW: 348.289 g/mol |
MDL Number:
MFCD00010045 CAS Number: 35193-64-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...