(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide, (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate

B1143-5G B1143-1G B1143-100MG
TCB1143-5GEA 232.26 CAD
TCB1143-5G TCB1143-001G TCB1143-100MG
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98.0% (by titrimetric analysis)
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

CAS Number: 39648-67-4
MDL Number: MFCD00010045
Molecular Formula: C20H13O4P
Molecular Weight: 348.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -595 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(-)-2,2'-Dihydroxy-1,1'-binaphthyl Hydrogen Phosphate

Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
MDL Number: MFCD00010045
CAS Number: 39648-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c32d1d21fc294232c1fa04a62e16e3f0f8fe16e0&vid=c32d1d21fc294232c1fa04a62e16e3f0f8fe16e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR