Lawesson's reagent ≥95.0% (by titrimetric analysis)
Supplier: TCI America
B1133-25G
B1133-250G
TCB1133-025GEA
64.29
CAD
TCB1133-025G
TCB1133-250G
Lawesson's reagent ≥95.0% (by titrimetric analysis)
Lawesson's reagent
[Sulfurating Reagent]
CAS Number: 19172-47-5
MDL Number: MFCD00005171
Molecular Formula: C14H14O2P2S4
Molecular Weight: 404.45
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 230
Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis)
Other Product Names / Synonyms: 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulfide
Formula:
C₁₄H₁₄O₂P₂S₄ Melting Pt: 230 °C |
MDL Number:
MFCD00005171 CAS Number: 19172-47-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...