Phenylglyoxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
B1129-5G
B1129-25G
TCB1129-005GEA
67.53
CAD
TCB1129-005G
TCB1129-025G
Phenylglyoxylic acid ≥97.0% (by GC, titration analysis)
Phenylglyoxylic acid
CAS Number: 611-73-4
MDL Number: MFCD00002575
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 138
Melting point (°C): 66
Other Product Names / Synonyms: Phenylglyoxylic Acid
Formula:
C₈H₆O₃ Boiling Pt: 138 °C Melting Pt: 66 °C |
MDL Number:
MFCD00002575 CAS Number: 611-73-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...