5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Bismuthiol II, 5-Mercapto-3-phenyl-1,3,4-thiadiazole-2-thione Potassium Salt

B1124-5G B1124-25G
TCB1124-005GEA 58.46 CAD
TCB1124-005G TCB1124-025G
5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt ≥98.0% (by HPLC, titration analysis)
5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt

CAS Number: 6336-51-2
MDL Number: MFCD00014092
Molecular Formula: C8H5KN2S3
Molecular Weight: 264.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 5-Mercapto-3-phenyl-1,3,4-thiadiazole-2-thione Potassium Salt

Formula: C₈H₅KN₂S₃
MDL Number: MFCD00014092
CAS Number: 6336-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb50c5ae8cd9fb8c42492beff47f1e8fe2129ddf&vid=cb50c5ae8cd9fb8c42492beff47f1e8fe2129ddf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR