(+)-cis-2-Benzylaminocyclohexanemethanol ≥98.0%

Supplier: TCI America

Synonyms: (+)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine, (1R,2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol

B1119-1G B1119-5G
TCB1119-1GEA 210.14 CAD
TCB1119-1G TCB1119-005G
(+)-cis-2-Benzylaminocyclohexanemethanol ≥98.0%
(+)-cis-2-Benzylaminocyclohexanemethanol

CAS Number: 71581-92-5
MDL Number: MFCD00151395
Molecular Formula: C14H21NO
Molecular Weight: 219.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 68
Specific rotation [a]20/D: 24 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: (+)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine

Formula: C₁₄H₂₁NO
MW: 219.33 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00151395
CAS Number: 71581-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b8b72e2108471c7841eb86ff9d03205d83dcca3&vid=2b8b72e2108471c7841eb86ff9d03205d83dcca3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR