(-)-cis-2-Benzylaminocyclohexanemethanol ≥98.0%
Supplier: TCI America
Synonyms:
(-)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine
B1118-5G
B1118-1G
TCB1118-005GEA
627.26
CAD
TCB1118-005G
TCB1118-1G
(-)-cis-2-Benzylaminocyclohexanemethanol ≥98.0%
(-)-cis-2-Benzylaminocyclohexanemethanol
CAS Number: 71581-93-6
MDL Number: MFCD00151396
Molecular Formula: C14H21NO
Molecular Weight: 219.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 68
Specific rotation [a]20/D: -24 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents
Other Product Names / Synonyms: (-)-N-Benzyl-cis-2-hydroxymethylcyclohexylamine
Formula:
C₁₄H₂₁NO MW: 219.33 g/mol Melting Pt: 68 °C |
MDL Number:
MFCD00151396 CAS Number: 71581-93-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...