2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 98,0
Supplier: TCI America
Synonyms:
2-Boc-thio-4,6-dimethylpyrimidine, O-tert-Butyl S-(4,6-dimethylpyrimidin-2-yl) carbonothioate
[Boc Agent for Peptides Synthesis]
CAS Number: 41840-28-2
MDL Number: MFCD00006080
Molecular Formula: C11H16N2O2S
Molecular Weight: 240.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 110
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: 2-Boc-thio-4,6-dimethylpyrimidine
Formula:
C₁₁H₁₆N₂O₂S MW: 240.32 g/mol Melting Pt: 51 °C Flash Pt: 110 °C |
MDL Number:
MFCD00006080 CAS Number: 41840-28-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...